
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7365-44-8
:
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532
:229.25 g/mol
:M-Clarity™ quality level = MQ200
EINECS:230-906-3
:GHS07
Storage Temperature:Ambient