trans-2,5-Dimethylpiperazine, Sigma-Aldrich®
Catalog # SIALD179604-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.19 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:2815-34-1
Formula:C₆H₁₄N₂
MDL Number:MFCD00070516