

Dibutyltin dilaurate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-58-7
:
Formula:C₃₂H₆₄O₄Sn
MDL Number:MFCD00008963
:631.57 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:201-039-8
UN:2788
:GHS06
:GHS09