TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)
Supplier: Roth Carl
Certificates
About this item
Synonyms: TAPS sodium salt, Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)propane-1-sulphonate
CAS Number:91000-53-2
:
Formula:C₇H₁₆NNaO₆S
MDL Number:MFCD00069903
Molecular Weight:265.26 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: