Order Entry
Export
ContactUsLinkComponent
 
1,1'-Bis(diphenylphosphino)ferrocene
Catalog # MOLE10332118-1G
Supplier:  Molekula
CAS Number:  
1,1'-Bis(diphenylphosphino)ferrocene
Catalog # MOLE10332118-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :554.39 g/mol
EINECS:430-420-3
CAS Number:12150-46-8
Formula:C₃₄H₂₈FeP₂
MDL Number:MFCD00001422