trans-2,5-Dimethylpiperazine
Catalog # MOLE57459985-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:114.19 g/mol
CAS Number:2815-34-1
Formula:C₆H₁₄N₂
MDL Number:MFCD00070516