2,6-Dimethylpiperazine
Catalog # MOLE41165860-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.19 g/mol
EINECS:203-588-9
UN:1325
ADR:4.1,II
CAS Number:108-49-6
Formula:C₆H₁₄N₂
MDL Number:MFCD00005956