Order Entry
Export
ContactUsLinkComponent
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Irritant
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Catalog # MOLEM10813371
Supplier:  Molekula
CAS Number:  
Irritant
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Catalog # MOLEM10813371
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :229.25 g/mol
EINECS:230-906-3
 :GHS07
Storage Temperature:Ambient
CAS Number:7365-44-8
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532