3,3',4,4'-Tetrachloroazoxybenzene
Catalog # EHERC17341000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:336 g/mol
CAS Number:21232-47-3
Formula:C₁₂H₆Cl₄N₂O