Order Entry
Export
Orders LinkContactUsLinkComponent
2-Fluoro-5-formylpyridine

2-Fluoro-5-formylpyridine

Glass bottle for solids, 5 g
Catalog # COBBOT-8140-5G
Supplier:   COMBI-BLOCKS
2-Fluoro-5-formylpyridine
Catalog # COBBOT-8140-5G
Supplier:  COMBI-BLOCKS
CAS Number:  677728-92-6
2-Fluoro-5-formylpyridine

Glass bottle for solids, 5 g

CAS Number:  677728-92-6
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  6-Fluoropyridine-3-carboxaldehyde, 6-Fluoronicotinaldehyde
  • CAS Number:677728-92-6
  • Formula:C₆H₄FNO
  • MDL Number:MFCD08064049
  • Molecular Weight:125.1 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications