Order Entry
Export
Orders LinkContactUsLinkComponent
1-Boc-4-piperidone

1-Boc-4-piperidone

Glass bottle for solids, 100 g
Catalog # COBBAM-1027-100G
Supplier:   COMBI-BLOCKS
1-Boc-4-piperidone
Catalog # COBBAM-1027-100G
Supplier:  COMBI-BLOCKS
CAS Number:  79099-07-3
1-Boc-4-piperidone

Glass bottle for solids, 100 g

CAS Number:  79099-07-3
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone
  • CAS Number:79099-07-3
  • Formula:C₁₀H₁₇NO₃
  • MDL Number:MFCD00151800
  • Molecular Weight:199.25 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications