Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3,5-dichlorobenzoate
Ethyl 3,5-dichlorobenzoate
Ethyl 3,5-dichlorobenzoate
Ethyl 3,5-dichlorobenzoate
Supplier:
Thermo Scientific
Certificates
Expand image
Ethyl 3,5-dichlorobenzoate
Supplier:
Thermo Scientific
About this item
CAS Number:
91085-56-2
:
Formula:
C₉H₈Cl₂O₂
MDL Number:
MFCD00221467
:
219.07 g/mol
EINECS:
252-373-6