Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,4-dichlorobenzoate
Methyl 2,4-dichlorobenzoate
Methyl 2,4-dichlorobenzoate
Methyl 2,4-dichlorobenzoate
Supplier:
Thermo Scientific
Certificates
Expand image
Methyl 2,4-dichlorobenzoate
Supplier:
Thermo Scientific
About this item
CAS Number:
35112-28-8
:
Formula:
C₈H₆Cl₂O₂
MDL Number:
MFCD00085117
:
205.04 g/mol
EINECS:
252-374-1