Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-3-nitrobenzonitrile
2,6-Difluoro-3-nitrobenzonitrile
2,6-Difluoro-3-nitrobenzonitrile
2,6-Difluoro-3-nitrobenzonitrile
Supplier:
Thermo Scientific
Certificates
Expand image
2,6-Difluoro-3-nitrobenzonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
143879-77-0
:
Formula:
C₇H₂F₂N₂O₂
MDL Number:
MFCD00042254
:
184.1 g/mol
UN:
3439
ADR:
6.1,III