
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:7365-44-8
:
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532
:229.25 g/mol
EINECS:230-906-3
:GHS07
Storage Temperature:Ambient