Order Entry
Export
ContactUsLinkComponent
Brij® 35 concentrate, high purity
Corrosive
Brij® 35 concentrate, high purity
Catalog # 0217-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Brij® 35 concentrate, high purity
Catalog # 0217-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Hydroxyl Number:
    40 – 60
  • Moisture (KF):
    ≤3%

Specification Test Results

About this item

 :
 :-
Melting Pt:39…43 °C
Flash Pt:149 °C
EINECS:500-002-6
 :GHS05
Storage Temperature:Ambient
CAS Number:9002-92-0
Formula:(C₂H₄O)nC₁₂H₂₆O
MDL Number:MFCD00080891