Order Entry
Export
ContactUsLinkComponent
Methyl-2-furoate 98%
Toxic
Methyl-2-furoate 98%
Catalog # ACRO156870250
CAS Number:  
undefined
Toxic
Methyl-2-furoate 98%
Catalog # ACRO156870250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Clear light yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4830 to 1.4890 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :126.11 g/mol
Boiling Pt:181 °C (760 mmHg)
Density:1.176 g/cm³
Flash Pt:73 °C
Storage Temperature:Ambient
EINECS:210-254-6
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:611-13-2
Formula:C₆H₆O₃
MDL Number:MFCD00003236