Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-2,5-Dimethylpiperazine
trans-2,5-Dimethylpiperazine
trans-2,5-Dimethylpiperazine
trans-2,5-Dimethylpiperazine
Supplier:
Thermo Scientific
Certificates
Expand image
trans-2,5-Dimethylpiperazine
Supplier:
Thermo Scientific
About this item
CAS Number:
2815-34-1
:
Formula:
C₆H₁₄N₂
MDL Number:
MFCD00070516
:
114.19 g/mol