Order Entry
Export
ContactUsLinkComponent
Cyclopentanol 99%
Flammable
Cyclopentanol 99%
Certificates
undefined
Flammable
Cyclopentanol 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4520 to 1.4540 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤50 APHA

Specification Test Results

Related Information