Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Cyclobutanedicarboxylic acid
1,1-Cyclobutanedicarboxylic acid ≥99%
1,1-Cyclobutanedicarboxylic acid ≥99%
:
Thermo Fisher Scientific
1,1-Cyclobutanedicarboxylic acid ≥99%
:
Thermo Fisher Scientific
:
5445-51-2
:
:
C₆H₈O₄
:
MFCD00001325
:
144.13 g/mol
:
663 °C (1013 hPa)
:
∼ 158 °C (dec.)
:
Ambient
:
226-651-2
:
3261
:
8,III
:
GHS05
msds_ALFAA14288
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years