1,1-Cyclobutanedicarboxylic acid 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Cyclobutane-1,1-dicarboxylic acid
- CAS Number:5445-51-2
- Formula:C₆H₈O₄
- MDL Number:MFCD00001325
- Molecular Weight:144.13 g/mol
- Boiling Pt:663 °C (1013 hPa)
- Melting Pt:157…161 °C
- Storage Temperature:Ambient
- EINECS:226-651-2
- UN:3261
- ADR:8,II
2 Options Available Below
Product Details & Documents
Synonyms: Cyclobutane-1,1-dicarboxylic acid
- CAS Number:5445-51-2
- Formula:C₆H₈O₄
- MDL Number:MFCD00001325
- Molecular Weight:144.13 g/mol
- Boiling Pt:663 °C (1013 hPa)
- Melting Pt:157…161 °C
- Storage Temperature:Ambient
- EINECS:226-651-2
- UN:3261
- ADR:8,II
Documents
ACRO11092
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:157 to 161 °C
- Water:≤0.1 % (K.F.)
- Loss on drying:≤0.1 % (50 °C)
- GC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 %
- Clarity of solution:(5 w/v% aq. soln.) clear



