1,2,5,6,9,10-Hexabromocyclododecane
Catalog # EHERC14152000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:641.7 g/mol
Boiling Pt:462 °C (1013 hPa)
Melting Pt:198…199 °C
EINECS:221-695-9
CAS Number:3194-55-6
Formula:C₁₂H₁₈Br₆