
2,4-Dimethylquinoline 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1198-37-4
:
Formula:C₁₁H₁₁N
MDL Number:MFCD00006760
:157.22 g/mol
Boiling Pt:264…265 °C
Density:1.060
Flash Pt:>110 °C (230 °F)
EINECS:214-832-9
:GHS07
Storage Temperature:Ambient