Order Entry
Export
ContactUsLinkComponent
4-Methylquinoline
4-Methylquinoline
Catalog # MOLEM16616281
Supplier:  Molekula
CAS Number:  
4-Methylquinoline
Catalog # MOLEM16616281
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Lepidine
 :
 :143.19 g/mol
Boiling Pt:261
EINECS:207-734-2
 :GHS07
Melting Pt:9
Storage Temperature:Ambient
CAS Number:491-35-0
Formula:C₁₀H₉N
MDL Number:MFCD00006784
Density:1.0995 g/cm³ (0 °C)
Merck Index:13,05460