4-Methylquinoline 99%
: Lepidine
:491-35-0
:
:C₁₀H₉N
:MFCD00006784
:261
:9…10 °C
:1.08 g/cm³
:143.19 g/mol
:>110 °C
:13,05460
:Ambient
:207-734-2
:GHS07
:GHS08
- Environmentally Preferable:
- Appearance:Clear light yellow to green to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6180 to 1.6220 (20°C, 589 nm)
- GC:≥98.5 %