

2,4-Quinolinediol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-95-3
:70254-43-2
Formula:C₉H₇NO₂
MDL Number:MFCD00006744
:161.16 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:>300 °C
EINECS:201-711-0
:GHS08
:GHS07
Storage Temperature:Ambient