8-Quinolinol
Glass bottle for solids, 1 kg
Catalog # MOLEM18897213
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 8-Hydroxyquinoline
- CAS Number:148-24-3
- Formula:C₉H₇NO
- MDL Number:MFCD00006807
- Molecular Weight:145.16 g/mol
- Merck Index:13,04869
- Boiling Pt:267 °C (1013 hPa)
- Melting Pt:71.2 °C
- Density:1.03 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-711-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



