Order Entry
Export
Orders LinkContactUsLinkComponent
8-Quinolinol ACS

8-Quinolinol ACS

Health
Irritant
8-Quinolinol ACS
Certificates
8-Quinolinol ACS
8-Quinolinol ACS

3 Options Available Below

Product Details & Documents

CAS No.: 148-24-3 Molecular Formula: C9H7NO Formula Weight: 145.16 Physical Form: Granular Sensitivity: Light Sensitive Appearance: White to pale yellow Melting Point: 72-76° Boiling Point: 267° Solubility: Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene. Beilstein Reference: 114512 Merck Reference: 14,4843 MDL No.: MFCD00006807

A bacteriostatic and fungistatic agent. Also a chelating agent for metals

Synonyms:  8-Hydroxyquinoline
  • CAS Number:148-24-3
  • Formula:C₉H₇NO
  • MDL Number:MFCD00006807
  • Molecular Weight:145.16 g/mol
  • Merck Index:14,4843
  • Boiling Pt:267 °C
  • Melting Pt:72…76 °C
  • Density:1.03 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-711-1
Product Family Options

Product Information