
5,7-Dichloro-8-quinolinol 98%
Catalog # ACRO113430050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to beige to grey Powder
- Infrared spectrum:Conforms
- Melting point:178 to 183 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:214.05 g/mol
Merck Index:13,02193
Boiling Pt:355 °C (1013 hPa)
Melting Pt:178…183 °C
EINECS:212-258-3
:GHS07
CAS Number:773-76-2
Formula:C₉H₅Cl₂NO
MDL Number:MFCD00006786