4,7-Dichloroquinoline ≥98%
Purity: 98% CAS No.: 86-98-6 Molecular Formula: C9H5Cl2N Formula Weight: 198.05 Flash Point: 164°(327°F) Melting Point: 83-87° Boiling Point: 148°/10mm Beilstein Reference: 125359 MDL No.: MFCD00006774
- :86-98-6
- :C₉H₅Cl₂N
- :MFCD00006774
- :198.05 g/mol
- :148 °C (10 mmHg)
- :83…87 °C
- :164 °C (327 °F)
- :Ambient
- :201-714-7
Purity: 98% CAS No.: 86-98-6 Molecular Formula: C9H5Cl2N Formula Weight: 198.05 Flash Point: 164°(327°F) Melting Point: 83-87° Boiling Point: 148°/10mm Beilstein Reference: 125359 MDL No.: MFCD00006774
- :86-98-6
- :C₉H₅Cl₂N
- :MFCD00006774
- :198.05 g/mol
- :148 °C (10 mmHg)
- :83…87 °C
- :164 °C (327 °F)
- :Ambient
- :201-714-7