
4,7-Dichloroquinoline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-98-6
:
Formula:C₉H₅Cl₂N
MDL Number:MFCD00006774
:198.05 g/mol
Boiling Pt:148 °C (760 mmHg)
Melting Pt:84…86 °C
Flash Pt:164 °C
Storage Temperature:Ambient
EINECS:201-714-7
:GHS07
Specification Test Results
- Appearance:White to yellow to brown Powder or crystals or crystalline solid
- Infrared spectrum:Conforms
- Melting point:84 to 88 °C
- GC:≥97.5 %