Quinaldic acid 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
- CAS Number:93-10-7
- Formula:C₁₀H₇NO₂
- MDL Number:MFCD00006752
- Molecular Weight:173.17 g/mol
- Merck Index:12,08227
- Melting Pt:159…161 °C
- Storage Temperature:Ambient
- EINECS:202-218-3
2 Options Available Below
Product Details & Documents
Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
- CAS Number:93-10-7
- Formula:C₁₀H₇NO₂
- MDL Number:MFCD00006752
- Molecular Weight:173.17 g/mol
- Merck Index:12,08227
- Melting Pt:159…161 °C
- Storage Temperature:Ambient
- EINECS:202-218-3
Documents
ACRO17105
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Light brown Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:157 to 159 °C
- Titration with NaOH:≥98.5 %



