Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Bromoquinoline
8-Bromoquinoline
8-Bromoquinoline
Irritant
8-Bromoquinoline
Supplier:
Thermo Scientific
Certificates
Expand image
Irritant
8-Bromoquinoline
Supplier:
Thermo Scientific
About this item
CAS Number:
16567-18-3
:
Formula:
C₉H₆BrN
MDL Number:
MFCD00191859
:
208.06 g/mol
:
GHS07