Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Phenylbenzaldehyde 99%

Irritant
4-Phenylbenzaldehyde 99%
Certificates
4-Phenylbenzaldehyde 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
  • CAS Number:3218-36-8
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00006947
  • Molecular Weight:182.22 g/mol
  • Boiling Pt:184 °C (11 mmHg)
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambient
  • EINECS:221-742-3

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    57 to 60 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,10 €
    29,10 €
    1 * 5 g
       29,10 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 84,00 €
    84,00 €
    1 * 25 g
       84,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 206,00 €
    206,00 €
    1 * 100 g
       206,00 €