2,6-Di-tert-butyl-p-cresol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:128-37-0
:
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644
:220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-881-4