p-Xylene
Catalog # EHERC17945200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:106.17 g/mol
Merck Index:12,10214
Boiling Pt:138.3 °C (1013 hPa)
Melting Pt:13 °C
Density:1.096 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:203-396-5
UN:1307
ADR:3,III
:GHS02
:GHS07
CAS Number:106-42-3
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556