Order Entry
Export
ContactUsLinkComponent
p-Xylene 99% for spectroscopy
Flammable
Irritant
Health
p-Xylene 99% for spectroscopy
Catalog # ACRO422670010
CAS Number:  
undefined
Flammable
Irritant
Health
p-Xylene 99% for spectroscopy
Catalog # ACRO422670010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥98.5 %
  • UV:
    (neat) 1 cm cell vs water
  • Color scale:
    ≤15 APHA
  • Suitability:
    (in liquid scint. counting) Passes test

Specification Test Results

About this item

 :
 :106.17 g/mol
Merck Index:12,10214
Boiling Pt:138.3 °C (1013 hPa)
Melting Pt:13 °C
Density:0.866 g/cm³
Flash Pt:25 °C
EINECS:203-396-5
UN:1307
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:106-42-3
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556