
1,4-Diisopropylbenzene 99%
:100-18-5
:
:C₁₂H₁₈
:MFCD00008892
:162.27 g/mol
:203 °C (760 mmHg)
:–17 °C
:0.86 g/cm³
:76 °C
:202-826-9
:GHS07
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4875 to 1.4905 (20°C, 589 nm)
- GC:≥98.5 %
- Chloride (Cl):18 to 19 %