Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diisopropylbenzene 96%

1,3-Diisopropylbenzene 96%

1,3-Diisopropylbenzene 96%
Certificates
1,3-Diisopropylbenzene 96%
1,3-Diisopropylbenzene 96%

2 Options Available Below

Product Details & Documents

Purity: 96% CAS No.: 99-62-7 Molecular Formula: C12H18 Formula Weight: 162.27 Flash Point: 76°(170°F) Refractive Index: 1.4890 Melting Point: -63° Boiling Point: 204° Beilstein Reference: 1905828 MDL No.: MFCD00008889

  • CAS Number:99-62-7
  • Formula:C₁₂H₁₈
  • MDL Number:MFCD00008889
  • Molecular Weight:162.27 g/mol
  • Boiling Pt:204 °C
  • Melting Pt:–63 °C
  • Density:0.856
  • Flash Pt:76 °C (170 °F)
  • Storage Temperature:Ambient
  • EINECS:202-773-1
  • UN:3082
  • ADR:9,III
Product Family Options

Product Information