Order Entry
Export
ContactUsLinkComponent
o-Xylene
Flammable
Irritant
o-Xylene
Catalog # PROOCHEUSC14
CAS Number:  
undefined
Flammable
Irritant
o-Xylene
Catalog # PROOCHEUSC14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :106.17 g/mol
Merck Index:12,10214
Boiling Pt:144 °C (1013 hPa)
Melting Pt:–25 °C
Density:0.88 g/cm³ (20 °C)
Flash Pt:30 °C (closed cup)
Storage Temperature:Ambient
EINECS:202-422-2
UN:1307
ADR:3,III
 :GHS02
 :GHS07
CAS Number:95-47-6
Formula:C₈H₁₀
MDL Number:MFCD00008519