Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Diethylbenzene
1,2-Diethylbenzene, Sigma-Aldrich®
1,2-Diethylbenzene, Sigma-Aldrich®
Flammable
Irritant
1,2-Diethylbenzene, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Flammable
Irritant
1,2-Diethylbenzene, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
135-01-3
:
Boiling Pt:
183 °C (1013 hPa)
Melting Pt:
–31.2 °C
Density:
0.8662 g/cm³ (18 °C)
EINECS:
205-170-1
UN:
2049
ADR:
3,III