2,4-Dimethylthiophenol 95%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 95% CAS No.: 13616-82-5 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 87°(189°F) Refractive Index: 1.5690 Odour: Unpleasant Boiling Point: 207-208° UN No.: 2810 MDL No.: MFCD00010019
Synonyms: 2,4-Dimethylbenzene-1-thiol, 1,3-Dimethyl-4-mercaptobenzene, 4-Mercapto-m-xylene, 2,4-Dimethylbenzenethiol
- CAS Number:13616-82-5
- Formula:C₈H₁₀S
- MDL Number:MFCD00010019
- Molecular Weight:138.23 g/mol
- Boiling Pt:207…208 °C
- Density:1.020
- Flash Pt:87 °C (189 °F)
- Storage Temperature:Ambient
- EINECS:237-100-0
- UN:2810
- ADR:6.1,III
- :Density: 1.027 g/cm³ (20 °C)
3 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 13616-82-5 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 87°(189°F) Refractive Index: 1.5690 Odour: Unpleasant Boiling Point: 207-208° UN No.: 2810 MDL No.: MFCD00010019
Synonyms: 2,4-Dimethylbenzene-1-thiol, 1,3-Dimethyl-4-mercaptobenzene, 4-Mercapto-m-xylene, 2,4-Dimethylbenzenethiol
- CAS Number:13616-82-5
- Formula:C₈H₁₀S
- MDL Number:MFCD00010019
- Molecular Weight:138.23 g/mol
- Boiling Pt:207…208 °C
- Density:1.020
- Flash Pt:87 °C (189 °F)
- Storage Temperature:Ambient
- EINECS:237-100-0
- UN:2810
- ADR:6.1,III
- :Density: 1.027 g/cm³ (20 °C)



