Order Entry
Export
ContactUsLinkComponent
3,4-Dimethylthiophenol
Irritant
3,4-Dimethylthiophenol
Catalog # MOLEM86280774
Supplier:  Molekula
CAS Number:  
Irritant
3,4-Dimethylthiophenol
Catalog # MOLEM86280774
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.23 g/mol
Boiling Pt:58…59 °C (1 mmHg)
Melting Pt:54…56 °C
Density:1.0256 g/cm³ (20 °C)
EINECS:242-587-8
UN:2810
ADR:6.1,III
 :GHS07
CAS Number:18800-53-8
Formula:C₈H₁₀S
MDL Number:MFCD00010023