Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Xylenol 99%

Toxic
Corrosive
Environment
2,4-Xylenol 99%
Certificates
2,4-Xylenol 99%

4 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-xylene, 2,4-Dimethylphenol, 2,4-Xylenol
  • CAS Number:105-67-9
  • Formula:C₈H₁₀O
  • MDL Number:MFCD00002233
  • Molecular Weight:122.17 g/mol
  • Merck Index:12,10215
  • Boiling Pt:211 °C
  • Melting Pt:21…23 °C
  • Density:1.027 g/cm³
  • Flash Pt:96 °C
  • Storage Temperature:Ambient
  • EINECS:203-321-6
  • UN:2928
  • ADR:6.1,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5375 to 1.5405 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information