


2,6-Xylenol 99%
:576-26-1
:
:C₈H₁₀O
:MFCD00002240
:122.17 g/mol
:12,10215
:203 °C (760 mmHg)
:45…48 °C
:1.15 g/cm³
:73 °C
:Ambient
:209-400-1
:2261
:6.1,II
:GHS06
:GHS05
:GHS09
- Appearance:Colorless to yellow-brown Low melting solid (May darken on storage)
- Infrared spectrum:Conforms
- Melting point:45 to 48 °C
- Water:≤0.05 %
- GC:≥98.5 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤3 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
- Iodine consuming subst.:≤0.004 meq/g