Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethylanisole ≥99%

3,5-Dimethylanisole ≥99%

3,5-Dimethylanisole ≥99%
Certificates
3,5-Dimethylanisole ≥99%
3,5-Dimethylanisole ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 874-63-5 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 65°(149°F) Refractive Index: 1.5130 Boiling Point: 192-193° Beilstein Reference: 2040905 MDL No.: MFCD00008398

Synonyms:  1-Methoxy-3,5-dimethylbenzene
  • CAS Number:874-63-5
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008398
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:192…193 °C
  • Melting Pt:–0.2 °C
  • Density:0.960
  • Flash Pt:65 °C (149 °F)
  • Storage Temperature:Ambient
  • EINECS:212-865-3
Product Family Options

Product Information