Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,6-Dimethylanisole
2,6-Dimethylanisole ≥98%
2,6-Dimethylanisole ≥98%
:
Thermo Fisher Scientific
2,6-Dimethylanisole ≥98%
:
Thermo Fisher Scientific
:
1004-66-6
:
:
C₉H₁₂O
:
MFCD00008380
:
136.19 g/mol
:
181…182 °C
:
0.962
:
67 °C (152 °F)
:
Ambient
:
213-723-3
msds_ALFAB20365
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years