2-Nitro-m-xylene
500 g
Catalog # MOLE65509553-500G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Nitro-m-xylene, 1,3-Dimethyl-2-nitrobenzene, 2,6-Dimethylnitrobenzene
- CAS Number:81-20-9
- Formula:C₈H₉NO₂
- MDL Number:MFCD00007181
- Molecular Weight:151.16 g/mol
- Boiling Pt:∼225 °C (1013 hPa)
- Melting Pt:14…16 °C
- Density:1.112 g/cm³ (20 °C)
- Flash Pt:87 °C
- Storage Temperature:Ambient
- EINECS:201-333-6
- UN:1665
- ADR:6.1,II
Specifications
Specifications
- Pk:500 g



