2-Nitro-m-xylene
Catalog # EHERC12728000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:151.16 g/mol
Boiling Pt:∼225 °C (1013 hPa)
Melting Pt:14…16 °C
Density:1.112 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:201-333-6
UN:1665
ADR:6.1,II
CAS Number:81-20-9
Formula:C₈H₉NO₂
MDL Number:MFCD00007181