Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Nitro-o-xylene

Supplier:   Molekula
3-Nitro-o-xylene
Supplier:  Molekula
Certificates
3-Nitro-o-xylene

2 Options Available Below

Product Details & Documents
Synonyms:  1,2-Dimethyl-3-nitrobenzene, 2,3-Dimethylnitrobenzene, 3-Nitro-o-xylene
  • CAS Number:83-41-0
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00007162
  • Molecular Weight:151.16 g/mol
  • Boiling Pt:245 °C (1013 hPa)
  • Density:1.1402 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-474-3
  • UN:1665
  • ADR:6.1,II
  •  :Density: 1.1402 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g